Whatman™ Quartz Air Sampling Filter Papers, Grade QM-C
Supplier: Cytiva
1855-045 1855-050
CA76498-680PK 433.84 CAD
CA76498-680 CA76498-682
Whatman™ Quartz Air Sampling Filter Papers, Grade QM-C
Filters Glass Fiber Filters
QM-C 100% quartz microfiber filter paper features low trace metals and high temperature resistance. It is well-suited for heavy metal analysis of particulate matter and sampling of acidic gases, stacks, flues and aerosols in air monitors.
  • 100% pure quartz microfiber, without any binder or organic chemicals
  • Pre-fired filters eliminate firing step, saving time in prepartion process
  • Air retention efficiency ≥99.95% at 0.3 μm DOP
QM-C high purity quartz microfiber filter addresses the challenge of heavy metal analysis in PM2.5/PM10. It is also suitable for organic chemical analyses such as PCBs and PAHs.
Certifications: Certified to conform to European and US EPA standards for heavy metal analysis in air particulate matter.