Cole-Parmer® Vials for Homogenizers
Supplier: SPEX SamplePrep
Cole-Parmer®
2256 2248 2241R-PEF 2310 2253-PC-48 2142-PE 2241-PEF-200 2258 2241-PC 2252-PC-30 2251-PC
76533-076PK 594 CAD
76533-076 76182-306 76533-078 76533-112 76182-308 76533-090 76182-310 76533-108 76182-300 76182-302 76533-084
Cole-Parmer® Vials for Homogenizers
Vials
Safely grind samples in the HG-600 Geno/Grinder® 2010 and HG-400 MiniG® homogenizers.
  • Screw thread cap ensures a tight seal
Cole-Parmer® vials are ideal for holding grinding balls and media. Vials are supplied with screw caps to secure and protect samples or grinding media.