PIP® NOVAX® Class 0 Rubber Insulating Sleeves
Supplier: PIP
193-0-LARGE 193-0-REGULAR 193-0-SMALL 193-0-XLARGE
76944-568PR 653.03 CAD
76944-568 76944-566 76944-564 76944-570
PIP® NOVAX® Class 0 Rubber Insulating Sleeves
Workwear
Natural rubber construction offers excellent dielectric properties combined with flexibility, strength, and durability.
  • Highest quality, natural latex with no solvents or additives
  • Strong, flexible, durable, and dielectric
  • Color coated label easily indicates compliance with regulatory standards
Ergonomically designed rubber sleeves feature a curved shape to help reduce fatigue. Straps and buttons help to keep the sleeve in position.
Environmentally friendly manufacturing process uses an aqueous dip process. This dipped construction process provides superior comfort and flexibility.
Certifications: Meets ASTM 496 and 1051 standards.