Assurance® Advanced Chemical Protection 7 mil Butyl Rubber Gloves, Rough or Smooth Grip
Supplier: PIP
1UB0007L 1UB0007M 1UB0007RM 1UB0007RS 1UB0007S 1UB0007X 1UB0007XS
76944-622CS 4508.83 CAD
76944-622 76944-620 76944-614 76944-612 76944-610 76944-618 76944-624 76944-616
Assurance® Advanced Chemical Protection 7 mil Butyl Rubber Gloves, Rough or Smooth Grip
Gloves
Butyl rubber gloves with a rough or smooth grip, available in 14" length.
  • Smooth finish
  • Ergonomic hand design
  • Easy to decontaminate
  • Excellent holdout
  • Made in the USA
Gloves protect against hazardous chemicals and materials.