Wards® The Ideal Gas Law
Supplier: ALDON CORP SE
Wards®
IS2528
470336-150EA 83.05 CAD
470336-150
Wards® The Ideal Gas Law
Educational Materials Chemistry Educational Materials Gas Analysis Learning Activities
Explore the gas laws through reaction of sodium bicarbonate and hydrochloric acid.
  • Kit contains enough materials for 15 groups of students teacher’s manual and student study guide copymasters are included
Explore the gas laws through the reaction of sodium bicarbonate and hydrochloric acid. Students will perform the reaction and use the ideal gas law to calculate the amount of carbon dioxide produced. They will then use stoichiometry to determine their percent yield
Delivery information: Includes 1×40 g Sodium bicarbonate; 15×25 ml Hydrochloric acid, 1 M; 1 pack String; 15 Balloons.