Heidolph® Hei-Dro Clean 165UC Glassware Washers
Supplier: Heidolph NA, LLC
023250040CA 023250041CA 023250042CA
CA77041-318EA 25917.39 CAD
CA77041-318 CA77041-320 CA77041-322
Heidolph® Hei-Dro Clean 165UC Glassware Washers
Glassware Washers
The Heidolph Tuttnauer Hei-Dro Clean 165UC glassware washers have an assortment of features and accessories to meet all laboratory washing needs. The Hei-Dro Clean is designed to sanitize, clean, and dry all labware such as beakers, bottles, graduated cylinders, ampoules, narrow-neck glassware and other glassware such as Erlenmeyer flasks, evaporation and receiving flasks, Soxhlet’s, condensers, Petri dishes, funnels, test tubes lids and stoppers.
  • Large viewing window of entire washing chamber
  • Lower spindle rack with 25 positions and 10 glassware holders
  • Upper standard rack; built-in water softener; purified water rinse
  • 20 preset programs (up to 20 user-created custom programs)
  • Spray arm monitoring
  • HEPA filter – HEPA-filtered forced air drying (through spindles with spindle rack)
  • RS232 Port for connection to an external printer
  • USB-B Port for connection to a PC
  • USB-A Port for on-board datalogging
The unit utilizes a color touch screen which provides easy operation and control of wash loads helping to ensure reproducible results. The large viewing window on the front of the unit allows users to closely monitor and view wash loads throughout a cycle. The Hei-Dro Clean 165UC glassware washer is supplied with an upper standard rack and a lower spindle rack with 25 positions and 10 glassware holders. Additional accessories and inserts are available to further customize the washer.
The units feature built-in water softener and purified water rinse. During a cycle, all wash parameters such as water temperature and number of stages, the active stage of the cycle, wash and rinse times, and cycle length are displayed on the display. Once a cycle is completed, HEPA-filtered forced air circulates throughout the chamber to dry the load. The unit features 20 preset programs and has the capacity to store up to 20 user-created custom programs. Different interfaces are available for on-board datalogging, connecting to a PC, and an external printer.
Certifications: CE marked.
Ordering information: Additional accessories, inserts and spindles are available to order separately.
Delivery information: Supplied with lower spindle rack and upper standard rack.