Dichloroacetic acid (DCA) 10% v/v toluene, BioSyn™ Detritylation Reagent, DNA/RNA synthesis, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: DCA Deblock, DCA

BR676-4X2.5L BR676ADAK-4
CABJBR67642.5LCS 1743.1 CAD
CABJBR67642.5L CABJBR676ADAK4
Dichloroacetic acid (DCA) 10% v/v toluene, BioSyn™ Detritylation Reagent, DNA/RNA synthesis, Burdick & Jackson™
Dichloroacetic acid (DCA)

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 194
Melting Pt: -4
Density: 0.9356 g/cm3 (20 °C)
Flash Pt: 8.33 °C
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
UN: 2924
ADR: 8 II
Merck Index: 13,03075

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR