Eisco Plastic Scoops
Supplier: Wards
CH0461APK12 CH0461BPK12 CH0461CPK12 CH0461EPK12 CH0461FPK6 CH0461GPK6 CH0461HPK6 CH0461IPK6
470343-956EA 23.95 CAD
470343-956 470343-970 470344-334 470344-582 470345-466 470345-894 470345-896 470346-052 470344-583 470343-957 470344-335 470345-895 470343-971 470346-053 470345-897 470345-467
Eisco Plastic Scoops
Scoops
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
  • Made of heat- and chemical-resistant polypropylene plastic
  • Scoop handles are labeled with capacity for easy identification
  • Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.