Anti-CD8a Mouse Monoclonal Antibody [SK1] (CF® 647 Plus)
Supplier: Biotium
P009-647P-125 P009-647P-500
77007-356EA 231.68 CAD
77007-356 77007-314
Anti-CD8a Mouse Monoclonal Antibody [SK1] (CF® 647 Plus)
Antibodies
CD8a (clone SK1) is a mouse monoclonal antibody derived from the CD8 alpha chain. This antibody belongs to the Biotium Choice list of select antibodies that have been validated and optimized in-house for optimal performance. The antibody is available conjugated to CF® Dyes and Astral™ tandem dyes. They are supplied in PBS, 0.1% BSA, 0.05% azide.
CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHCMore Product Information