Rapid Response™ Fentanyl Test Strips (Liquid/Powder)
Supplier: BTNX INC.
FYL-18S7-100 FYL-18S7-100
76945-146EA 157.78 CAD
76945-146 CA76945-146
Rapid Response™ Fentanyl Test Strips (Liquid/Powder)
Drug Tests
The Rapid Response™ Fentanyl test strips (Liquid/Powder) detects fentanyl in suspicious substances on surfaces and liquids from suspicious receptables at a cut-off concentration of 200 ng/ml.
  • Sample: Liquid/Powder
  • Detects fentanyl at a cut-off concentration of 200 ng/ml
  • Results interpretation guide printed on each pouch
  • Easy to interpret results with built-in procedural controls
  • Individually packaged strips
  • Sensitive to fentanyl and analogues: Carfentanil, Butyryl Fentanyl, p-Fluoro Fentanyl, Acetyl Fentanyl, Fentanyl, Furanyl Fentanyl, Valeryl Fentanyl, Ocfentanil, 3-Methyl Fentanyl, Remifentanil, Sufentanil
Warning: This test does not evaluate drug safety or purity.
The Rapid Response™ Fentanyl test strip (Liquid/Powder) detects the presence of fentanyl in liquid and powder substances at a cut-off of 200 ng/ml or higher.
It is specific for fentanyl screening with no significant cross-reactivity to other opiates, such as morphine and heroin. This kit includes individually packaged test strips for sanitary with result interpretations printed on the back of each pouch, a result interpretation card to aid in reading the results, and detailed instructions.
To perform the test, remove the test strip from the pouch and hold the strip at the end where the product name is printed. Dip the test strip into liquid or powder sample (diluted in water) for 10 to 15 seconds. Remove the strip from the sample and place it on a non-absorbent flat surface. A negative result can be interpreted as soon as both the test (T) and control (C) lines appear. A positive result can be interpreted once the control line has appeared and the background has cleared to white, typically by 60 seconds. Do not read results after ten minutes. Please read the Product Insert supplied with the kit before use.
Caution: The test device is not intended to determine the purity, composition, or if the substance being examined is safe to use.
A positive or negative test result is NOT an indication that the substance being examined is safe to use. Many factors come into play when examining the samples, including but not limited to mixture of multiple substances, solubility, and pH of the sample.
BTNX Inc. does not encourage the use, supply, or production of illegal drugs or controlled substances in any way. The device is intended for harm reduction purposes. Follow the advice of your local harm reduction or public health agency.
There is a possibility that technical or procedural errors as well as other substances and factors may interfere with the Rapid ResponseTM Fentanyl Test Strip (Urine) and cause false results.
A positive result indicates the presence of fentanyl only and does not indicate quantity.
A negative result does not at any time rule out the presence of fentanyl, as they may be present below the minimum detection level of the test.