Masterflex® Tri-Clamp to Hose Barb Adapters, 316SS
Supplier: VWR International
MFLX30510-10 MFLX30510-11 MFLX30510-12 MFLX30510-13 MFLX30510-14 MFLX30510-15 MFLX30510-16 MFLX30510-17 MFLX30510-18 MFLX30510-19 MFLX30510-20 MFLX30510-21 MFLX30510-22 MFLX30510-23 MFLX30510-24 MFLX30510-25 MFLX30510-26 MFLX30510-27 MFLX30510-28 MFLX30510-29 MFLX30510-00 MFLX30510-01 MFLX30510-02 MFLX30510-03 MFLX30510-04 MFLX30510-05 MFLX30510-06 MFLX30510-07 MFLX30510-08 MFLX30510-09 MFLX30510-40 MFLX30510-41 MFLX30510-30 MFLX30510-31 MFLX30510-32 MFLX30510-33 MFLX30510-34 MFLX30510-35 MFLX30510-36 MFLX30510-37 MFLX30510-38 MFLX30510-39
MFLX30510-10EA 183.28 CAD
MFLX30510-10 MFLX30510-11 MFLX30510-12 MFLX30510-13 MFLX30510-14 MFLX30510-15 MFLX30510-16 MFLX30510-17 MFLX30510-18 MFLX30510-19 MFLX30510-20 MFLX30510-21 MFLX30510-22 MFLX30510-23 MFLX30510-24 MFLX30510-25 MFLX30510-26 MFLX30510-27 MFLX30510-28 MFLX30510-29 MFLX30510-00 MFLX30510-01 MFLX30510-02 MFLX30510-03 MFLX30510-04 MFLX30510-05 MFLX30510-06 MFLX30510-07 MFLX30510-08 MFLX30510-09 MFLX30510-40 MFLX30510-41 MFLX30510-30 MFLX30510-31 MFLX30510-32 MFLX30510-33 MFLX30510-34 MFLX30510-35 MFLX30510-36 MFLX30510-37 MFLX30510-38 MFLX30510-39
Masterflex® Tri-Clamp to Hose Barb Adapters, 316SS
Fittings and Connectors
High-quality tri-clamp to hose barb fittings in 316 stainless steel for sanitary applications.
  • Constructed from 316 stainless steel for excellent corrosion resistance and durability
  • Designed to meet sanitary standards, ensuring hygienic fluid transfer in critical applications
  • Secure tri-clamp design allows for reliable attachment to various equipment and systems
  • Tailored hose barb shape and size options to accommodate a wide range of hose sizes
  • Precision engineered and designed for use with tubing clamps to ensure tight seals for leak-free connections, maintaining product integrity
  • Suitable for various industries including food and beverages, pharmaceuticals, and more
  • Capable of handling demanding operating conditions with resistance to high temperatures and pressures
  • Designed to work seamlessly with existing equipment, enhancing overall system compatibility
Elevate your fluid transfer systems with these premier sanitary tri-clamp to hose barb fittings in 316 stainless steel. Engineered for excellence, these fittings combine top-grade materials with hygienic design, ensuring unparalleled durability, corrosion resistance, and leak-free performance. Their versatile compatibility with various hose types and sizes, seamless tri-clamp attachment, and easy maintenance make installation a breeze, and supporting industries ranging from food and beverage to pharmaceuticals. Trust in these fittings to enhance operational efficiency, promote system integrity, and set new benchmarks for fluid management excellence.