Eisco Plastic Büchner Funnels, 2 Part
Supplier: Wards
CH0464C CH0464A CH0464B
470344-032EA 17 CAD
470344-032 470345-474 470345-760
Eisco Plastic Büchner Funnels, 2 Part
Funnels Filtration Funnels Büchner Funnels
Two-part Büchner funnels with straight sides and fixed perforated plates made of polypropylene plastic.
  • For use in filtration (separation of solid matter and liquid)
  • Made of high-quality, chemically-resistant polypropylene which is lightweight and autoclavable
  • Features straight sides and a perforated filter plate on top part
  • Two interlocking parts form a vacuum seal which does not require greasing
  • Filtration paper required but not included
  • Often used in combination with a Büchner flask, Büchner ring, and sinter seals
Büchner funnels with straight sides and fixed perforated plates are made of lightweight, autoclavable polypropylene. Designed with two interlocking parts which form a vacuum seal that does not require greasing. For use in filtration (separation of solid matter and fluid). Filtration paper required but not included. Moisten filter paper with a liquid to prevent initial leakage.