SCIEX Triple Quad 6500+ LC-Systems
Supplier: AB SCIEX LLC CA
5062192 5062195 5062192 5062195
76644-986EA 744453.39 CAD
76644-986 76644-988 CA76644-986 CA76644-988
SCIEX Triple Quad 6500+ LC-Systems
Spectrophotometers and Photometers Mass Spectrometers
The Triple Quad 6500+ system features multi-component IonDrive technology that pushes the boundaries of quantitation. The sensitivity, speed, and performance delivered through these technology enhancements enable you to see it all, from low mass to high mass compounds.
  • QTRAP Activated and QTRAP ready options
  • Source: IonDrive Turbo V ion source with TurboIonSpray probe or APCI probe
  • Increased throughput with polarity switching speeds of 5 ms
  • No compromise in mass range with a 2000 m/z upper mass limit
  • Scan speeds of up to 12000 Da/s for optimized UHPLC strategies
  • QTRAP activated (optional): Triple quadrupole: 5 to 2000 Da; linear ion trap: 50 to 2000 Da
  • 5 ms in MRM and scheduled MRM modes
Superior signal-to-noise and LOQ for the widest scope of chemical compounds in the most challenging matrices. IonDrive high energy detector (QTRAP activated). Enhanced ion transmission for added selectivity without compromising LOQs. Compatible with optional SelexION+ differential mobility spectometry. Up to six orders of linear dynamic range for an enhanced breadth of applications.
The Triple Quad 6500+ system delivers revolutionary sensitivity, speed, and performance for your most challenging methods.
The Triple Quad 6500+ system, QTRAP activated option offers revolutionary sensitivity, speed, and performance for your most challenging methods. This system features the multi-component IonDrive™ technology, including the IonDrive™ turbo V source and newly designed IonDrive high energy detector, and is ideal for laboratories requiring the ultimate in performance. Options include Electrospray Ionization (ESI) and heated nebulizer ionization probes. This system is ideal for laboratories performing demanding applications requiring ultimate sensitivity for ultra-low-level quantitation of small and large molecules.