EXP®2 Fitting Kits for Stainless Steel Tubing
Supplier: Optimize
15-20-04933 15-19-04962 15-19-04959 15-19-04961 15-19-04958 15-20-04924
10798-584PK 437.31 CAD
10798-584 10798-586 10798-588 10798-776 10798-778 10798-780
EXP®2 Fitting Kits for Stainless Steel Tubing
Fittings and Connectors
Optimize is proud to introduce EXP2 - the next generation of EXP UHPLC fitting technology. Featuring a small ³/₁₆" hex-head EXP2 nut, the new EXP2 Driver and the original titanium hybrid ferrule, the EXP2 fittings offer hand-tight connections rated to 20000 psi+. The EXP2 nuts are available with 6-32, 6-40 and 10-32 threads and have been scaled down to allow for maximum working room in cramped spaces. The EXP2 Driver is a small, easy to use knurled torque driver which snaps on to the nuts for hand-tightening and loosening. The EXP2 Driver is slotted and can be removed from the nut and tubingMore Product Information