OPTI-PEEK™ Tubings
Supplier: Optimize
10-13-00168 10-13-00170 10-13-00177 10-13-00179 10-13-00181 10-13-00183 10-13-00186 10-13-00187 10-13-00191 10-13-00175
77100-126EA 298.37 CAD
77100-126 77100-128 77100-132 77100-134 77100-136 77100-138 77100-140 77100-142 77100-144 10123-222
OPTI-PEEK™ Tubings
Tubing Chromatography Tubing
Biocompatible and chemically inert to most commonly-used solvents tubing.
  • Can withstand pressures up to 6000 psi
  • Alternative to stainless steel tubing for high pressure applications
  • Withstands continuous use at HPLC pressure without swelling or bursting
  • Not affected by aggressive mobile phases
  • Will not corrode