OPTI-TRAP™ Holders
Supplier: Optimize
10-02-04745 10-02-04747 10-02-04749 10-02-04751
10122-678EA 1488.45 CAD
10122-678 10122-680 10122-682 10122-684
OPTI-TRAP™ Holders
Chromatography Supplies Chromatography Spare Parts
The OPTI-TRAP™ is a bi-directional trap cartridge system which is used individually or in a series for sample concentration and purification.
  • For use up to 1500psi (100bar)
  • Low back pressure, for syringe loading/eluting or on-line trapping applications when plumbed into sample injection valve
  • Titanium and CTFE for Chemical and Biocompatibility
  • Bidirectional for easy flushing and regeneration of the bed
  • Three bed sizes: 50 μl, 5.0 μl and 0.5 μl
The OPTI-TRAP system features a durable biocompatible trap holder assembly made of either PEEK (capillary configuration) or stainless steel with a PEEK flow path (micro and macro configurations). OPTI-TRAPs feature low back pressure and are loaded and eluted manually with a syringe or on-line when plumbed into the sample injection valve. OPTI-TRAPs include titanium frits for full biocompatibility.
Ordering information: Custom packing available.