Nexus™ Horizontal Clean Bench, 8' Wide
Supplier: Labconco
5380710 5380800 5380810 5381000 5381010 5381100 5381110 5381200 5381210 5381300 5380110 5380200 5380210 5380300 5380310 5380500 5380510 5380600 5380610 5380700 5381310 5381500 5381510 5381600 5381610 5381700 5381710 5381800 5381810 5380000 5380010 5380100
77038-018EA 29711.9 CAD
77038-018 77038-020 77038-022 77038-024 77038-026 77038-028 77038-030 77038-032 77038-034 77038-036 77037-998 77038-000 77038-002 77038-004 77038-006 77038-008 77038-010 77038-012 77038-014 77038-016 77038-038 77038-040 77038-042 77038-044 77038-046 77038-048 77038-050 77038-052 77038-054 77037-992 77037-994 77037-996
Nexus™ Horizontal Clean Bench, 8' Wide
Fume Hoods and Enclosures Laminar Flow Cabinets
Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area.
  • Efficient unidirectional laminar airflow provided through CAP™ technology
  • Intelligent 5" touchscreen interface features intuitive monitoring capabilities
  • Reliable with long-life HEPA filters and maximum cleanability that stands up to disinfectants
The Nexus Horizontal clean bench is a laminar flow workstation designed to protect non-hazardous samples and processes from contamination.
The Nexus is the most advanced clean bench onMore Product Information