BioX Cryogenic Vials, Barcoded
Supplier: TWD Scientific
BX88-6051 BX88-6052 BX88-6053 BX88-6101 BX88-6102 BX88-6103 BX88-6151 BX88-6152 BX88-6153 BX88-9051 BX88-9052 BX88-9053 BX88-9151 BX88-9152 BX88-9153 BX88-6201 BX88-6202 BX88-6203 BX88-6502 BX88-6503 BX88-6214 BX88-6215 BX88-6500 BX88-6504 BX88-6505 BX88-9050 BX88-9054 BX88-9055 BX88-9100 BX88-9101 BX88-6150 BX88-6154 BX88-6155 BX88-6200 BX88-6204 BX88-6205 BX88-6210 BX88-6211 BX88-6212 BX88-6213 BX88-9501 BX88-9502 BX88-9503 BX88-9504 BX88-9505 BX88-9102 BX88-9103 BX88-9104 BX88-9105 BX88-9150 BX88-9154 BX88-9155 BX88-9200 BX88-9201 BX88-9202 BX88-6050 BX88-6054 BX88-6055 BX88-6100 BX88-6104 BX88-6105 BX88-9203 BX88-9204 BX88-9205 BX88-9210 BX88-9211 BX88-9212 BX88-9213 BX88-9214 BX88-9215 BX88-9500
76404-402CS 538.1 CAD
76404-402 76404-404 76404-406 76404-408 76404-410 76404-412 76404-414 76404-416 76404-418 76404-420 76404-422 76404-424 76404-426 76404-428 76404-430 76404-432 76404-434 76404-436 76404-440 76404-442 77395-002 77395-004 77395-006 77395-008 77395-010 77395-012 77395-014 77395-016 77395-018 77395-020 77394-982 77394-984 77394-986 77394-988 77394-990 77394-992 77394-994 77394-996 77394-998 77395-000 77395-062 77395-064 77395-066 77395-068 77395-070 77395-022 77395-024 77395-026 77395-028 77395-030 77395-032 77395-034 77395-036 77395-038 77395-040 77394-970 77394-972 77394-974 77394-976 77394-978 77394-980 77395-042 77395-044 77395-046 77395-048 77395-050 77395-052 77395-054 77395-056 77395-058 77395-060
BioX Cryogenic Vials, Barcoded
Vials
Barcoded polypropylene cryogenic vials with colored caps.
  • Suitable for liquid nitrogen storage
  • Pre-barcoded
  • O-ring free
  • Self standing
  • White writing patch and graduations
Unique permanent barcodes for superior traceability. 1D and human-readable on the side are perfect for hand scanning and the 2D barcodes on the bottom make these vials automation friendly.
Caution: Use in liquid phase LN₂ is not recommended.