Anti-SARS-CoV-2 Nucleocapsid Rabbit Monoclonal Antibody [clone: N66]

Supplier: Genscript
GenScript®
A02137-100 A02137-1
77007-538EA 637.75 CAD
77007-538 77007-626
Anti-SARS-CoV-2 Nucleocapsid Rabbit Monoclonal Antibody [clone: N66]
Antibodies
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease 2019 (COVID-19). Nucleocapsid Protein is a most abundant structure protein of the coronavirus which is associated with nucleic acid.

  • High specificity
  • High affinity
  • High stability

Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards, and control antibodies.

Type:
Antigen: SARS-CoV-2 Nucleocapsid
Clonality: Monoclonal
Clone: N66
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR