Anti-Panitumumab Mouse Monoclonal Antibody [clone: 9G12]

Supplier: Genscript
GenScript®
A02100-40
77007-492EA 243.16 CAD
77007-492
Anti-Panitumumab Mouse Monoclonal Antibody [clone: 9G12]
Antibodies
Panitumumab with trade name Vectibix, is an FDA-approved drug for the treatment of patients with EGFR-expressing metastatic colorectal carcinoma with non-mutated (wild-type) KRAS after failure of fluoropyrimidine-, oxaliplatin- and irinotecan-containing chemotherapy regimens. It is a human monoclonal antibody. Panitumumab binds to the human EGF receptor (EGFR), preventing binding of activating ligands, such as the EGF and transforming growth factor alpha. Anti-Panitumumab antibody (9G12), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with panitumumab.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Panitumumab
Clonality: Monoclonal
Clone: 9G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR