Sodium chloride solution 2 M, in single-use containers, HyClone™ for bioprocessing

Supplier: Cytiva

SH31141.01 SH31141.02 SH31141.03 SH31141.04 SH31141.05 SH31141.06
CA76894-100EA 265.18 CAD
CA76894-100 CA76894-122 CA76894-120 CA76894-132 CA76894-168 CA76894-172
Sodium chloride solution 2 M, in single-use containers, HyClone™ for bioprocessing
Sodium chloride

HyClone™ NaCl solutions in single-use bags offer a convenient format for many upstream and downstream bioprocessing applications.


  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.2 µm sterile filtered.
  • Provided in single-use bag containers.
  • Raw material is EP, USP, and JP grade.


These products are manufactured upon order (100 L minimum order quantity). HyClone™ NaCl in single-use bags is well-suited for use with: Cytiva's chromatography resins.
Available in 1 M and 2 M sodium chloride concentrations up to 200 L. Contact us for information around bag configurations or custom bulk formulations.

Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Specification Test Results

Storage Room Temperature as defined by regional Pharmacopeia (15-30°C in US; 15-25°C in EU)
Endotoxin Concentration ≤ 1 EU/mL
Certifications/Compliance Manufactured in a ISO9001 and ISO13485 certified facility
Shelf life 12 Months
For use with Chromatography resins, IC systems, ÄKTA hardware systems
Appearance Clear, colorless solution
Sterility No Growth

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR