NOVAX® Class 00 Insulating Gloves with Straight Cuff, Orange
Supplier: PIP
147-00-11/10 147-00-11/11 147-00-11/12 147-00-11/6
76944-320PR 100.61 CAD
76944-320 76944-322 76944-324 76944-318
NOVAX® Class 00 Insulating Gloves with Straight Cuff, Orange
Gloves
Rubber Insulating gloves are critical PPE for EV manufacturing. They help protect automotive workers against electrical shock if contact is made to an energized surface.
  • Strong, flexible, durable, and dielectric
Critical PPE for EV manufacturing. Gloves should meet and/or exceed the requirements of the ASTM D120-14a Standard specification for rubber insulating gloves. Gloves should also be electrically tested following ASTM D120-14a.
Certifications: Gloves should meet and/or exceed the requirements of the ASTM D120-14a.