VSV-Pseudovirus_HCV H77 Strain Luciferase
Supplier: ReVacc Scientific
This pseudotyped virus uses recombinant vesicular stomatitis virus (rVSV) to carry the full length E1E2 of Hepatitis C virus (HCV) (H77 strain) (GenBank: AAB67036.1).
- Anti-Hepatitis C virus neutralizing antibody screening at high throughput
- Anti-Hepatitis C virus drug screening at high throughput
- Hepatitis C virus vaccine efficacy evaluation at high throughput
- Hepatitis C virus pseudovirus transduction of target cells for viral entry, receptor recognition, cellular tropism and functional studies, such as ADCC analysis.
Description: Infection of cells with this pseudotyped virus carrying luciferase reporter results in high level luciferase activity. See our titration result showing that the starting 2-fold diluted pseudovirus generates signal ~ 100,000-fold higher than uninfected control (cell alone as background in blue).We also evaluated the neutralizing activity of one positive antibody RV36 by using this pseudotyped virus.
Size: 200 μl with recommended 500-fold dilution. We recommend to use it at a dilution fold where the signal of pseudovirus infection is 1,000-fold higher than uninfected control (cell alone as background), although 100-1, 000-fold high is acceptable. Can be used for 1,000 reactions (100 μl diluted virus) or 2,500 reactions if using reduced amount of pseudovirus (40 μl diluted virus).
Use protocol: Incubate 100 μl diluted pseudovirus with 100 μl of your sample in each well of 96-well plate with at least one duplicate for 30 to 60 min at 37 °C. Then add 100 μl target cells (Huh-7 or others expressing receptor). Read luciferase signal next day. For protocol using reduced amount, incubate 40 μl diluted virus with 10 μl your sample in each well of 96-well plate for 30 to 60 min at 37 °C. Then add 25 μl target cells. Add 150 μl fresh media next day and read signal on Day 3. To be noted for neutralizing titer (such as EC50) calculation, more duplication leads to more accurate reading.
Packaging: Shipping with dry ice. Require −80 °C storage. Multiple freeze/Thaw cycles will reduce its sensitivity. Recommend only one cycle. Aliquot after the first thaw.
Caution: Handle it in biosafety cabinet in BSL-2. Contacted tips and tubes should be decontaminated by 10% disinfecting bleach. Pseudoviruses are intended for research use only.