Eisco Mortars and Pestles
Supplier: Wards
CH0540D CH0540F CH0540C CH0540B CH0540G CH0540A CH0540E
470343-716EA 19.6 CAD
470343-716 470343-850 470345-006 470345-806 470345-822 470346-068 470346-082
Eisco Mortars and Pestles
Mortars and Pestles Mortars with Pestles
Mortar and Pestle sets made of solid porcelain with spout and unglazed grinding surfaces.
  • Mortar and pestle are both made of solid porcelain
  • Resistant to chips and cracks
  • Inside grinding surface of mortar and the rounded end of the pestle are unglazed for precise grinding and crushing
  • Mortar features spout for easy pouring and transferring of crushed material
Single Mortar and Pestle set perfect for finely grinding and crushing grains, pigments, powders, pills, and more. Mortar and pestle are both made of solid porcelain and are resistant to chips and cracks. Inside grinding surface of mortar and the rounded end of the pestle are unglazed for precise grinding and crushing. Mortar features a contoured spout for pouring and transferring of crushed material.