Eisco Dropping Funnels with PTFE Stopcock
Supplier: Wards
CH0474D CH0474A CH0474E CH0474B CH0474C
470343-976EA 60.75 CAD
470343-976 470344-250 470344-952 470345-210 470345-588
Eisco Dropping Funnels with PTFE Stopcock
Funnels Dropping Funnels
Dropping funnels with PTFE stopcocks and sockets fitted with interchangeable plastic stoppers.
  • Cylindrical ungraduated glass dropping funnel
  • Fitted with an interchangeable plastic stopper and PTFE key stopcock
  • Made of durable, high quality borosilicate 3.3 glass which withstands high temperatures and thermal shocks
  • Used for extractions or to dispense liquids into a system slowly (drop-wise)
Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco Labs Borosilicate glassMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable