VWR® Thermal-Transfer Cryogenic Labels, Zebra-Compatible
Supplier: VWR International
76808-842 76808-870 76808-878 76808-840 76808-966 76808-972 76808-864 76808-962 76808-948 76808-952 76808-852 76808-960 76808-828 76808-900 76808-912
76808-842RL 30.47 CAD
76808-842 76808-870 76808-878 76808-840 76808-966 76808-972 76808-864 76808-962 76808-948 76808-952 76808-852 76808-960 76808-828 76808-900 76808-912
VWR® Thermal-Transfer Cryogenic Labels, Zebra-Compatible
Labels
Thermal Transfer Cryogenic Labels are ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications.
  • Can be placed in most incubators
  • Labels are thermal transfer top coated and require a ribbon for printing
Labels feature a unique adhesive that can withstand extreme temperatures from –196 to 121 °C (–321 to 250 °F), without the need for lamination.
Ordering information: Rolls of labels come on a 1 or 3" core, depending on the intended printer model.
Order Now
SPECIFICATIONS
Environmentally Preferable