VWR® Silicone Sealing Mats for VWR® Deep Well Plates
Supplier: SCIENTIFIC SPECIALTIES, INC.
76M1-00-131AV 76M1-S0-131AV 76924-060 77M1-S0-131AV
76924-052CS 291.53 CAD
76924-052 76924-054 76924-060 76924-062
VWR® Silicone Sealing Mats for VWR® Deep Well Plates
Seals Microplate Seals
Provide a safe, secure seal to minimize sample loss with VWR® Deep Well Storage Plates.
  • Reusable and pierceable
  • For use in pharmaceutical industry, food industry, and molecular biology
  • Temperature range: –80 to +120 °C
These mats are compatible with VWR® Deep Well Plates in the series: 76924-044 through 76924-058.
Certifications: Certified free of DNase, RNase, DNA and pyrogens. Sterile mats are also endotoxin-free.