ep Dualfilter T.I.P.S.® SealMax® Filter Pipette Tips, BioBased Reloads
Supplier: Eppendorf
0030081234 0030081242 0030081250 0030081269 0030081277
CA76881-956PK 311.74 CAD
CA76881-956 CA76881-958 CA76881-960 CA76881-962 CA76881-964
ep Dualfilter T.I.P.S.® SealMax® Filter Pipette Tips, BioBased Reloads
Pipette Tips
Eppendorf ep Dualfilter T.I.P.S.® SealMax® BioBased Reloads in 'Biopur®' grade blend eco-innovation with ultimate purity. Made from 90% plant-based material in a reloadable format, these self-sealing, dual-filter tips are ideal for sensitive, contamination-free lab applications.
  • Improve the laboratory carbon footprint with ep Dualfilter T.I.P.S.® SealMax® BioBased in the new Eppendorf Reloads
  • Produced from recycled feedstock in second generation, derived solely from waste and residue streams from vegetable oil production
  • Significantly reduce laboratory
More Product Information