Eppendorf twin.tec® Trace PCR Plates, 96 Wells
Supplier: Eppendorf
0030129784 0030129768 0030129776
CA76881-970PK 196.05 CAD
CA76881-970 CA76881-966 CA76881-968
Eppendorf twin.tec® Trace PCR Plates, 96 Wells
Microplates PCR Plates
The twin.tec® Trace PCR Plates have improved ergonomics and traceability. These microtiter plates are the perfect choice for high-throughput applications, automated workflows, and standardised processes.
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • A new innovative optical guiding grid for quick well-orientation simplifies your manual and semi-automated liquid handling steps
  • Fuchsia and crystal blue colours support better visibility and contrast
  • Customized barcode versions are available
The new twin.tec Trace plates are comparable in terms of physical material, specifications and superior reproducibility to the well-established twin.tec plates.
Available in transparent and 2 fresh colours twin.tec Trace plates are ready to deliver maximum performance and reliability for your most demanding workflows.