SteraMist SteraPak
Supplier: Tomi Environmental Solutions
S-PAK
76919-428EA 14136.96 CAD
76919-428
SteraMist SteraPak
Disinfection Systems
The all-in-one SteraPak places the power of SteraMist onto your back, delivering innovative disinfection that utilizes a rechargeable battery, cordless operation, and a new compact BIT solution bottle. 
  • The fully portable back-mounted SteraMist disinfection unit that offers unprecedented flexibility
  • Easily switch between battery and direct power
  • Delivers ionized Hydrogen Peroxide (iHP) in the most lightweight SteraMist system
  • For use with BIT-100 Solution
Perfect for maneuvering through small areas, such as attics and crawlspaces.
Delivery information: Includes SteraMist handheld applicator with 4’ cord, 1× SteraPak battery and charger, 32 oz DI water cartridge.