PHCbi VIP® ECO SMART Ultra-Low Temperature Upright Freezers –80 °C
Supplier: PHC Corporation
MDF-DU503VHA-PA MDF-DU703VHA-PA
CA76637-148EA 38051.19 CAD
CA76637-148 CA76637-152
PHCbi VIP® ECO SMART Ultra-Low Temperature Upright Freezers –80 °C
Freezers Ultra Low Temperature Freezers
These ULT freezers are purpose-built for reliable, long-term sample storage and designed to achieve and maintain temperature uniformity and stability. The VIP ECO SMART series combine next generation engineering with SMART control technology to provide ULT freezers with excellent energy efficiency and enhanced security and operation. Proven frost mitigation technology lowers the total cost of ownership.
  • Controlled freezer access − Screen lock with password protection, facial recognition
  • LCD touchscreen
  • 3 access ports − Upper wall (back-up system), bottom left (recorder sensor), bottom right (access) 17 mm (0.6")
  • Next generation engineering harnesses the power of inverter compressor technology to yield greater temperature uniformity
  • Category-leading energy efficiency (VIP ECO SMART)
  • Enhanced frost mitigation (FrostLess)
  • Temperature uniformity throughout the chamber
  • Innovative security control features
ULT technology innovation in action. The VIP® ECO SMART features next-generation engineering to deliver industry-leading energy efficiency and enhanced security. It uses 30% less energy than other leading ENERGY STAR® Certified freezers*. The FrostLess offers advances that mitigate frost buildup while still delivering energy efficiency.
These FrostLess ULT freezers offer less frost build-up than category-leading models, while maintaining the reliability and energy efficiency today's laboratories require. Their new thermal insulation design helps prevent large amounts of frost buildup on the outside of the inner doors, reducing maintenance requirements. *Based upon www.energystar.gov/productfinder/product/certified-lab-grade-refrigeration/ website accessed Jan 11, 2023
Certifications: Energy Star certified.
Delivery information: All units are supplied with a five-year manufacturer's parts and labor warranty.