Adjustable Sampling Tubes for Bioreactors, M10
Supplier: Chemglass
CLS-1380-21 CLS-1383-08 CLS-1385-13
10170-320EA 503.78 CAD
10170-320 10170-322 75780-150
Adjustable Sampling Tubes for Bioreactors, M10
Tubes Test Tubes
These sampling tubes are adjustable to permit the culture fluid to be sampled at any desired level.
Tubing is ¹/₄" OD. Stainless steel components manufactured from 316 L with an electropolished surface finish of 20Ra or better.