AccuForge® Single Edge Blades, Unwrapped
Supplier: AccuTec Blades
AccuForge®
AGBL-7043-0000 AGBL-7026-0000 AGBL-7016-0000
76717-310CS 1918.69 CAD
76717-310 76717-336 76717-324
AccuForge® Single Edge Blades, Unwrapped
Razors and Blades
AccuForge blades are unwrapped and paper free to reduce particulates.
  • Paper free blades are clean room ready
  • Bulk, unwrapped blades are ideal for high volume use
  • Choose 3-facet blades for increased sharpness, or 2-facet blades for increased durability
  • Available in 0.012" heavy duty carbon steel
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.