AccuForge® Single Edge Blades, Unwrapped
Supplier: Accutec Blades
AGBL-7022-0000 AGBL-7050-0000
76717-320CS 6696.64 CAD
76717-320 76717-340
AccuForge® Single Edge Blades, Unwrapped
Razors and Blades
AccuForge blades are unwrapped and paper free to reduce particulates.
  • Available with MicroCoat® enrichment technology
  • Re-closable clamshell for safe and convenient blade storage
  • Degreased to remove oil residue
  • Paper-free blades are clean room ready
  • Choose 3-facet blades for increased sharpness, or 2-facet carbon blades for increased durability
  • Each clamshell contains 115 blades
AccuForge's MicroCoat® blade enrichment technology adds a layer of protection for each blade helping maintain a sharp effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.