VWR® UHP Multichannel Pipettes
Supplier: VWR International
76627-800 76627-780 76627-788 76627-768 76627-794 76627-778 76627-782 76627-774
76627-800EA 667.61 CAD
76627-800 76627-780 76627-788 76627-768 76627-794 76627-778 76627-782 76627-774
VWR® UHP Multichannel Pipettes
Pipettes Micropipettes
The VWR® UHP multichannel pipette is available in sizes from 10 to 300 µl with a lightweight and durable design. These pipettes will provide you with low pipetting forces and the highest levels of accuracy and precision.
  • Autolocking volume mechanism to avoid volume drift while pipetting
  • Fully autoclaveable and UV resistant
  • Color coded ejector button and calibration switch make for easy identification
  • Universal fit with most common brands of pipette tips
  • Lightweight design, quality materials, low pipetting forces, and a 5-ear warranty
The VWR® multichannel pipette is calibrated to ISO 8655 standards using VWR® Universal pipette tips. Pipettes were tested for both autoclave and UV resistance.
Certifications: In compliance with ISO 8655.
Accessories information: Pipette Stands are available in the accessories tab below.
Packaging: Includes a QC certificate, pipette, and a quick reference guide.