miniPCR® BioBits® @home Central Dogma
Supplier: AMPLYUS LLC
KT-1102-01 KT-1102-10
470352-044EA 204.39 CAD
470352-044 470352-000
miniPCR® BioBits® @home Central Dogma
Educational Classroom Kits and Activities
Bring cutting-edge synthetic biology right into any learning setting!
The BioBits® Central Dogma @home version includes all the reagents and hardware you need to explore gene expression in any learning setting!
BioBits® pellets are tiny molecular factories that can create a variety of proteins, from brightly colored fluorescent proteins to functional enzymes - no fancy equipment or complicated protocols needed. Researchers have been using cell-free reactions in their laboratories for years, and now the BioBits® cell-free system makes this cutting-edge technology accessible anywhere to anyone interested in learning molecular biology. In this activity, you will use BioBits® cell-free technology to visualize the flow of genetic information and monitor transcription and translation in real time through fluorescence. Learn about the central dogma of molecular biology with cutting-edge synthetic biology - even in remote learning settings!
Shipping and storage notes:
- These reagents ship overnight.
- Store in a freezer (–20 °C) and use within 6 months of receipt.
- If a freezer is not available, viable for three months when stored in a fridge (4 °C ).
Delivery information: Includes BioBits® Central Dogma mini-lab (enough reagents to do the activity twice); P51™ Molecular Fluorescence Viewer; 9-volt battery; 4 μl minipette (fixed-volume micropipette); 20 disposable micropipette tips; activity guide.