miniPCR® Lab Starter Pack
Supplier: AMPLYUS LLC
QP-2500-08 QP-2516-16
470352-064EA 2496.29 CAD
470352-064 470352-092
miniPCR® Lab Starter Pack
Educational Classroom Kits and Activities
The essential tools of molecular biology in one handy bundle!
Delivery information: This bundle includes the following:miniPCR® mini8X or mini16X thermal cycler: a powerful, portable PCR machine always at your fingertips; blueGel™ system: all-in-one electrophoresis and transillumination; Set of 3 micropipettes (1 to 10 μl, 2 to 20 μl, 20 to 200 μl): Universal fit, compatible with standard micropipette tips, and easy to re-calibrate; Consumables starter pack: all the tubes, tips, and gel electrophoresis reagents you need to hit the ground running. Includes agarose (electrophoresis grade, 20More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable