HABITAT Lid Stands for HABITAT research Bioreactors
Supplier: IKA Works
35004059 35004067
76836-718EA 3007.44 CAD
76836-718 76836-682
HABITAT Lid Stands for HABITAT research Bioreactors
Bioreactors
This sturdy lid stand can be used for 2, 5 and 10 L glass vessels.
You can conveniently hang the lid on the stand, so that the individual components can be easily mounted or dismounted. In addition, you have the option to put the motor and sensors in designated places, so that the workplace remains tidy. The pressure spring tripod rod also makes it easy to move the tripod up and down.