HyClone™ CDM4CHO, CHO Cell Culture Platform Liquid Media, Cytiva
Supplier: Cytiva
SH30557.02 SH30558.02 SH30556.01
CA82013-294EA 146.72 CAD
CA82013-294 CA82013-300 CA76635-876
HyClone™ CDM4CHO, CHO Cell Culture Platform Liquid Media, Cytiva
Cell Culture Media
HyQ® CDM4CHO™ is a chemically-defined protein-free medium containing no animal derived components. It is designed to increase the process yields for the industrial manufacture of recombinant proteins using a variety of CHO cell clones. HyQ® CDM4CHO™ has been successfully tested in various culture systems, including T-flasks, shaker flasks and bioreactors. HyQ® CDM4CHO™ contains Pluronic® F68 and L-glutamine, and does not contain phenol red.
  • Metabolically designed for high cell yield and recombinant protein production
  • Allows for direct or sequential adaptation
  • Designed for large-scale culture applications, including perfusion and fed-batch strategies
  • All components fully traceable
  • Manufactured under cGMP
HyClone CDM4CHO contains Pluronic F68 and L-glutamine, and does not contain phenol red. It is also available without L-glutamine to support the GS gene expression system.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature 2...8 °C