HyClone™ SFM4CHO media, Cytiva
Supplier: Cytiva
SH30548.02 SH30549.02 SH30518.01
CA82013-380EA 110.57 CAD
CA82013-380 CA82013-386 CA76635-858
HyClone™ SFM4CHO media, Cytiva
Cell Culture Media
HyClone™ SFM4CHO media is a serum-free and protein-free formulation that contains no components of bovine origin and has been designed for high performance in a variety of culture vessels, including bioreactors.
  • Metabolically designed for high cell yield and recombinant protein production
  • Supports multiple CHO clones
  • Allows for direct or sequential adaptation
  • Supports growth in multiple cell culture systems
  • Component traceability
  • Manufactured under cGMP
HyClone™ SFM4CHO media has been formulated using a proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines. It has been designed to support the DHFR selection/amplification system and is also available without L-glutamine to support the GS gene expression system.