Syringe filters, Acrodisc®, Supor® membrane, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
4659 4661 4662 4602 4508 4608 4612 4651 AP-4426 4504 4655 4525 4506 4604 4611 4614 4618 4652 4654 AP-4425 4658 4187 4656 4653 4692 4509 4187 4668 4650 AP-4799 4692
CA28139-710PK 3767 CAD
CA28139-710 CA28143-044 CA28143-046 CA28143-048 CA28143-300 CA28143-303 CA28143-304 CA28143-310 CA28143-348 CA28143-374 CA34181-078 CA34181-088 CA28143-295 CA28143-301 CA28143-302 CA28143-309 CA28143-312 CA28143-314 CA28143-350 CA28143-352 CA28143-372 CA28143-400 CA28144-009 CA28150-958 CA34181-086 97035-174 28143-305 97027-952 28144-009 CA97035-176 CA28150-956 CA97027-954 CA97035-174 CA77603-480
Syringe filters, Acrodisc®, Supor® membrane, Cytiva (Formerly Pall Lab)
Filters Syringe Filters
Acrodisc™ syringe filters with Supor™ membrane deliver high flow rates and low protein binding.
  • Superior flow rates and higher throughputs than competitive devices
  • Low protein binding to minimize sample loss
  • Pre-sterilized products are sterilized by gamma irradiation to eliminate potential contamination by EtO residuals
  • A range of sizes (13 to 37 mm) accommodates sample volumes from < 10 to 150 ml
  • Acrodisc PF and Serum Acrodisc syringe filters feature built-in prefilter for increased throughput of difficult-to-filter liquids
  • Reduces mycoplasma with the use of 0,1 μm pore size
Supor™ hydrophilic polyethersulfone (PES) membrane: PP housing (except 25 and 32 mm: Acrylic and Acrodisc Serum: ABS)
Applications:
0.1 and 0.2 μm pore sizes provide sterilization of small volumes of buffers, culture media, and additives.
Acrodisc PF and Serum Acrodisc syringe filters are ideal for clarification/sterilization of viscous or particulate-laden solutions.
Use larger pore size filters for prefiltration and particulate removal.
Certifications: Passes United States Pharmacopeia (USP) Biological Reactivity Test, In Vivo <88>.