ACCU-SCOPE® ioLight Ultra-Portable Digital Microscopes

Supplier: ACCU-SCOPE INC SE
IOL-100-XY IOL-200 IOL-200-XY
470351-160EA 4530.15 CAD
470351-160 470351-216 470351-098
ACCU-SCOPE® ioLight Ultra-Portable Digital Microscopes
Microscopes
Compact and rechargeable digital microscopes deliver live HD images directly to a mobile device, ideal for field study and education.

  • Field of view 1 mm or 2 mm
  • Magnification of 400× or 150×
  • Working distance 1 mm or 2.5 mm
  • LED Illumination
  • Stage 1 mm and 2 mm glass or XY mechanical stage
  • Can display on smart phone, tablet or computer via Wi-Fi
  • 5 MP color camera
  • Brightfield, phase contrast or fluorescence
  • Integrated Li-ion rechargeable battery

With top and bottom illumination and accepting standard glass microscope slides and coverslips, ioLight portable digital microscopes have a variety of applications. Control the microscope, view/capture/share images with the mobile device app or an internet browser. Available with 1 mm or 2 mm field of view, with and without XY positioning control.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR