RSA-Pro X™ Surface treated vial and cap kits
Supplier: MicroSolv Technology Corp
9512S-3DCP-PX 9512S-3DMP-PX 9512S-KDAP-PX 9512S-KDCP-PX 9512S-KDMP-PX 9509S-1DAP-PX 9509S-1DCP-PX 9509S-3DAP-PX 9509S-3DCP-PX 9509S-KDAP-PX 9509S-KDCP-PX 9512S-1DAP-PX 9512S-1DCP-PX 9512S-1DMP-PX 9512S-3DAP-PX
76881-728PK 490.76 CAD
76881-728 76881-730 76881-734 76881-736 76881-738 76881-708 76881-710 76881-712 76881-714 76881-716 76881-718 76881-720 76881-722 76881-724 76881-726
RSA-Pro X™ Surface treated vial and cap kits
Vials
Due to the propriety coating process these vials and inserts have extreme hydrolytic stability making them stable in water and other solvents for a very long period of time.
  • Prevent adsorption of hydrophilic compounds that normally adhere to borosilicate glass vials
  • A totally hydrophobic surface coverage out performs silanizing vials when deactivation is needed
These vials and inserts can be autoclaved or frozen. Chosen when silanizing is not satisfactory or you want to improve your sample recovery.