dNTP sets, illustra™, Cytiva
Supplier: Cytiva
28-4065-51 28-4065-52 28-4065-53 28-4065-57 28-4065-58
CA95038-254LEA 734.19 CAD
CA95038-254L CA95038-256L CA95038-258L CA95038-260L CA95038-262L
dNTP sets, illustra™, Cytiva
Nucleic Acid Reagents Nucleotides
High purity dNTPs for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for consistent, high performance
  • Buffer-free, ready to use solutions at a variety of concentrations
  • Functionally tested to produce a 20,7 kb PCR amplification product from lambda DNA