UVP Gel Compact Bioimaging Systems
Supplier: Analytik Jena CA
849-97-0947-01 849-97-0947-02 849-97-0947-03 849-97-0947-04
CA76837-118EA 8229.7 CAD
CA76837-118 CA76837-098 CA76837-110 CA76837-112
UVP Gel Compact Bioimaging Systems
Photodocumentation Systems
The UVP Gel Compact imager is a cellphone-driven bioimaging system designed for the documentation and analyzes of various types of samples which include DNA gels, protein gels, colony plates and more. Capture images and analyze samples via your personal mobile device equipped with Analytik Jena's VisionWorks Software App.
  • All-in-one, compact gel documentation
  • Use the camera on your phone for stunning gel images
  • Cellphone-driven VisionWorks software app
  • Epi-white and epi-blue LED illumination to enable a variety of gel imaging applications
The UVP Gel Compact imager is an autonomous, independent, and cellphone-driven bioimaging system designed for the documentation and analyzes for various types of samples which include but are not limited to DNA gels, protein gels, and colony plates and more. The UVP Gel Compact allows users to capture images and analyze them via their personal mobile device equipped with Analytik Jena's VisionWorks Software App. The new VisionWorks application was generated for easy gel capture and analyzes and allows for total control of the UVP Gel Compact darkroom.
The system features a 302 nm wavelength transilluminator, overhead white and blue LEDs, 4 position automated emission filter tray, and phone holder/adapter that can accommodate most phone sizes on the market.
Enjoy all the capabilities of our VisionWorks® Touch software on your smartphone by using our VisionWorks® App (available for iOS and Android). Our VisionWorks® App is a powerful software that allows the capture of gel images and analyzes through your cellphone while having full control of the darkroom. This feature makes the UVP Gel Compact, the perfect match for the educational market.
Key features of the VisionWorks App include: User friendly interface (self-explanatory icons), software controlled lighting and filter (connection through Bluetooth), auto and manual focus in the software, auto and manual capture, automatically save the captured image in the phone's gallery, cloud analyzes for 1D lanes and bands, auto detection, molecular weight calibration, area density analyzes, colony counting and classification, result report viewable in the App, report shareable via email.