Autoclave Bags with Steam Indicator, Self-Seal, Heavy Duty SMS
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BGSMS3SS2126 BGSMS3SS2424 BGSMS3SS2430 BGSMS3SS2435 BGSMS3SS2630 BGSMS3SS2836 BGSMS3SS3030 BGSMS3SS3636 BGSMS3SS3648 BGSMS3SS4848 BGSMS3SS09.516 BGSMS3SS1217 BGSMS3SS1221 BGSMS3SS1318 BGSMS3SS14.522 BGSMS3SS1520 BGSMS3SS1622 BGSMS3SS1824 BGSMS3SS1830 BGSMS3SS2028
76809-154CS 2281.13 CAD
76809-154 76809-162 76809-164 76809-170 76809-134 76809-140 76809-144 76809-148 76809-150 76809-136 76809-146 76809-152 76809-158 76809-168 76809-138 76809-166 76809-142 76809-156 76809-172 76809-160
Autoclave Bags with Steam Indicator, Self-Seal, Heavy Duty SMS
Bags Sterilization Pouches
Self Sealing Heavy Duty SMS Autoclave Bags from Keystone Cleanroom Products are designed to satisfy the most stringent requirements in the sterilization of critical pharmaceutical equipment and components.
  • Convenient, time saving self-seal closure system
  • Significant improvement versus cutting and wrapping with sheets and tape
  • Superior barrier prevents liquid and microbial penetration
  • Manufactured and packaged in a certified ISO 6 Cleanroom
  • Strong, rip and tear resistant 50 gsm SMS material
  • Sub packed into sealed labeled poly bags with manufacturing lot
More Product Information