VSV-Pseudovirus_SARS-CoV-2 Omicron BA.2.12.1 Strain Spike with Luciferase Reporter
Supplier: ReVacc Scientific
This pseudotyped virus uses recombinant vesicular stomatitis virus (rVSV) to carry the S protein of SARS-CoV-2 (GenBank: MN908947) with multiple mutations initially identified in variant of Omicron BA.2.12.1. The S has 18-aa cytoplasmic tail truncation for optimal infection.
- Anti-SARS-CoV-2 neutralizing antibody screening at high throughput
- Anti-SARS-CoV-2 drug screening at high throughput
- COVID-19 vaccine efficacy evaluation at high throughput
- SARS-CoV-2 pseudovirus transduction of target cells for viral entry, receptor recognition, cellular tropism and functional studies, such as ADCC analysis
Infection of cells with this pseudotyped virus carrying luciferase reporter results in high level luciferase activity. See our titration result showing that the starting 2-fold diluted pseudovirus generates signal over 100000-fold higher than uninfected control (cell alone as background in blue). We also evaluated the neutralizing activity of one own S2-specific antibody RV6 and one published antibody COVA1-16 by using this pseudotyped virus.
Size: 200 μl with recommended 250-fold dilution. We recommend to use it at a dilution fold where the signal of pseudovirus infection is 1000-fold higher than uninfected control (cell alone as background), although 100 to 1000-fold high is acceptable. Can be used for 500 reactions (100 μl diluted virus) or 1250 reactions if using reduced amount of pseudovirus (40 μl diluted virus).
Use protocol: Download complete protocols from our website. Briefly, incubate 100 μl diluted virus with 100 μl your sample in each well of 96-well plate with at least one duplicate for 30 to 60 min at 37 °C. Then add 100 μl target cells (Vero E6 or others expressing ACE2 receptor). Read luciferase signal next day. For protocol using reduced amount, incubate 40 μl diluted virus with 10 μl your sample in each well of 96-well plate for 30 to 60 min at 37 °C. Then add 25 μl target cells. Add 150 μl fresh media next day and read signal on Day 3.
Accessories information: See suggested protocol to use this product.
Packaging: Shipping with dry ice. Require −80 °C storage. Multiple freeze/thaw cycles will reduce its sensitivity. Recommend only 1 cycle. Aliquot after the first thaw.
Caution: Handle it in biosafety cabinet in BSL-2. Contacted tips and tubes should be decontaminated by 10% disinfecting bleach. Pseudoviruses are intended for research use only.