Anti-His Tag Mouse Monoclonal Antibody [clone: mRV7]

Supplier: ReVacc Scientific
R44J3-100UG R44J3-500UG
76729-592EA 522.89 CAD
76729-592 76729-466
Anti-His Tag Mouse Monoclonal Antibody [clone: mRV7]
Antibodies
This antibody has been tested to bind to His tagged protein.

Type: Primary
Antigen: His Tag
Clonality: Monoclonal
Clone: mRV7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR