Virus Recombinant Monkeypox Dimeric A35R Protein (from Insect cell), Myc and His Tags
Supplier: ReVacc Scientific
S95L46-100UG S95L46-500UG
76729-510EA 606.58 CAD
76729-510 76729-646
Virus Recombinant Monkeypox Dimeric A35R Protein (from Insect cell), Myc and His Tags
Proteins and Peptides
A recombinant form of monkeypox virus protein A35R (MPXV_USA_2022_MA001; ON838178.1, isolate SI2022_S7) was produced by insect cells, followed by purification. This product contains ectodomain only (58-end), N-terminal Myc tag and C-terminal 6 hexa-histidine tag. The ectodomain includes cysteines for dimer formation. The dimeric conformation was evaluated with SEC (Superose 6 column) with two peaks that we assigned one as a dimer fraction peak. The product is not a SEC fraction.
This product has been run on SDS-PAGE with reduced condition. We also evaluated its binding to antibodies of anti-His, anti-Myc or clone RV10.