Virus Recombinant Monkeypox Trimeric A29 Protein (from Insect cell), His Tag
Supplier: ReVacc Scientific
A recombinant form of monkeypox virus protein A29 ( MPXV_USA_2022_MA001; ON838178.1, isolate SI2022_S7) was produced by insect cells, followed by purification. This product contains trimerization domain and C-terminal 6 hexa-histidine tag. The trimeric conformation was evaluated with SEC (Superose 6 column) with two peaks that we assigned one as a trimer fraction peak. The product is not a SEC fraction.
This product has been run on SDS-PAGE with reduced condition. We also evaluated its binding to anti-His antibody with a negative control.